ENA413166402 Enamine

1-(1-tert-butyl-4-hydroxy-1H-pyrazol-3-yl)ethan-1-one

  • MDL number MFCD17926206
  • Cas Number : 1197815-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141519
Availability details for
Quantity
  • Availability
  • Add to Cart