ENA413166405 Enamine

2-(cyclopropylmethoxy)ethan-1-ol

  • MDL number MFCD16102647
  • Cas Number : 56988-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140344
Availability details for
Quantity
  • Availability
  • Add to Cart