ENA413166407 Enamine

1-nitro-2-(prop-2-yn-1-yloxy)benzene

  • MDL number MFCD09030813
  • Cas Number : 13350-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139748
Availability details for
Quantity
  • Availability
  • Add to Cart