ENA413166416 Enamine

2-methyl-2H-indazole-3-carboxylic acid

  • MDL number MFCD07368556
  • Cas Number : 34252-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224...226
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141608

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141608
Availability details for
Quantity
  • Availability
  • Add to Cart