ENA413166419 Enamine

5-bromo-4-methylthiophene-3-carboxylic acid

  • MDL number MFCD24107252
  • Cas Number : 1523182-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171...173
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141612

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141612
Availability details for
Quantity
  • Availability
  • Add to Cart