ENA413166421 Enamine

4-iodobut-1-ene

  • MDL number MFCD11848508
  • Cas Number : 7766-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141614

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141614
Availability details for
Quantity
  • Availability
  • Add to Cart