ENA413166422 Enamine

3-(propan-2-yloxy)cyclobutan-1-amine

  • MDL number MFCD21657748
  • Cas Number : 1488811-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141616

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141616
Availability details for
Quantity
  • Availability
  • Add to Cart