ENA413166428 Enamine

methyl 2-sulfamoylpyridine-4-carboxylate

  • MDL number MFCD25966976
  • Cas Number : 75903-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141626

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141626
Availability details for
Quantity
  • Availability
  • Add to Cart