ENA413166431 Enamine

4-amino-2-cyclopentylpyrimidine-5-carbonitrile

  • MDL number MFCD19678702
  • Cas Number : 17247-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167...169
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140365

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140365
Availability details for
Quantity
  • Availability
  • Add to Cart