ENA413166432 Enamine

ethyl 2,4,4-trimethylpent-2-enoate

  • MDL number MFCD26895512
  • Cas Number : 1807938-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141631

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141631
Availability details for
Quantity
  • Availability
  • Add to Cart