ENA413166436 Enamine

3-tert-butyl-[1,2,4]triazolo[4,3-a]pyridine-6-carboxylic acid

  • MDL number MFCD07376857
  • Cas Number : 1250541-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 299...301
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140367

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140367
Availability details for
Quantity
  • Availability
  • Add to Cart