ENA413166437 Enamine

methyl 3-aminothieno[3,2-c]pyridine-2-carboxylate

  • MDL number MFCD08234760
  • Cas Number : 111042-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225-227
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141641
Availability details for
Quantity
  • Availability
  • Add to Cart