ENA413166438 Enamine

2-methoxy-3-methylbutanoic acid

  • MDL number MFCD09947964
  • Cas Number : 66018-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141642
Availability details for
Quantity
  • Availability
  • Add to Cart