ENA413166446 Enamine

(2-phenyloxolan-3-yl)methanol

  • MDL number MFCD21864559
  • Cas Number : 1525526-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140368
Availability details for
Quantity
  • Availability
  • Add to Cart