ENA413166453 Enamine

7-bromo-2,3-dimethyl-1H-indole

  • MDL number MFCD21193805
  • Cas Number : 877160-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141694
Availability details for
Quantity
  • Availability
  • Add to Cart