ENA413166454 Enamine

1-methyl-2-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD19228580
  • Cas Number : 30932-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141695

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141695
Availability details for
Quantity
  • Availability
  • Add to Cart