ENA413166457 Enamine

2-[(2-methylbenzenesulfonyl)methyl]benzoic acid

  • MDL number MFCD26936206
  • Cas Number : 1184738-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141701

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141701
Availability details for
Quantity
  • Availability
  • Add to Cart