ENA413166460 Enamine

6-oxo-1-{[2-(propan-2-yl)-1,3-thiazol-4-yl]methyl}-1,6-dihydropyridine-3-carboxylic acid

  • MDL number MFCD16404135
  • Cas Number : 1282763-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153...155
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140371

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140371
Availability details for
Quantity
  • Availability
  • Add to Cart