ENA413166465 Enamine

3-{[5-(4-nitrophenyl)furan-2-yl]methylidene}-2,3-dihydro-1H-indol-2-one

  • MDL number MFCD03937407
  • Cas Number : 64258-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141711

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141711
Availability details for
Quantity
  • Availability
  • Add to Cart