ENA413166466 Enamine

2-[(2,6-difluorophenyl)formamido]acetic acid

  • MDL number MFCD00218151
  • Cas Number : 66648-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141712
Availability details for
Quantity
  • Availability
  • Add to Cart