ENA413166468 Enamine

2-(2-bromophenoxy)pyrazine

  • MDL number MFCD09939178
  • Cas Number : 129242-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141714
Availability details for
Quantity
  • Availability
  • Add to Cart