ENA413166469 Enamine

4-[(3-methylbutanamido)methyl]benzoic acid

  • MDL number MFCD08722964
  • Cas Number : 925580-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141715
Availability details for
Quantity
  • Availability
  • Add to Cart