ENA413166470 Enamine

{2-amino-1-[3-(trifluoromethyl)phenyl]ethyl}dimethylamine

  • MDL number MFCD09735130
  • Cas Number : 96788-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140372
Availability details for
Quantity
  • Availability
  • Add to Cart