ENA413166478 Enamine

2H,3H,6H,7H,8H-[1,4]dioxino[2,3-f]indol-7-one

  • MDL number MFCD12403673
  • Cas Number : 1216180-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141730
Availability details for
Quantity
  • Availability
  • Add to Cart