ENA413166480 Enamine

2-(2,2-diethoxyethoxy)benzaldehyde

  • MDL number MFCD21364936
  • Cas Number : 23145-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141739
Availability details for
Quantity
  • Availability
  • Add to Cart