ENA413166483 Enamine

1-[3-(1-methylcyclopropyl)-1H-pyrazol-5-yl]piperazine dihydrochloride

  • MDL number MFCD26936213

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141746
Availability details for
Quantity
  • Availability
  • Add to Cart