ENA413166484 Enamine

2-[4-(bromomethyl)phenyl]-2-methylpropanenitrile

  • MDL number MFCD20184913
  • Cas Number : 1201643-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141751

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-141751
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart