ENA413166494 Enamine

2-(ethoxycarbonyl)-3,3-dimethylbutanoic acid

  • MDL number MFCD20638324
  • Cas Number : 83096-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141768

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141768
Availability details for
Quantity
  • Availability
  • Add to Cart