ENA413166495 Enamine

[6-(morpholin-4-yl)pyridin-2-yl]methanamine

  • MDL number MFCD08435862
  • Cas Number : 868755-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141769

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141769
Availability details for
Quantity
  • Availability
  • Add to Cart