ENA413166502 Enamine

2-methyl-4-[(pyrrolidin-3-yloxy)methyl]-1,3-thiazole dihydrochloride

  • Molecular Weight : 271.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-4-((pyrrolidin-3-yloxy)methyl)thiazole dihydrochloride
Inchi Code 1S/C9H14N2OS.2ClH/c1-7-11-8(6-13-7)5-12-9-2-3-10-4-9;;/h6,9-10H,2-5H2,1H3;2*1H
Inchi Key HVXACERJKCZANO-UHFFFAOYSA-N

Description

Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-141784

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141784
Availability details for
Quantity
  • Availability
  • Add to Cart