ENA413166515 Enamine

2-chloro-5-(dimethylsulfamoyl)benzene-1-sulfonyl chloride

  • MDL number MFCD09729704
  • Cas Number : 98491-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126...128
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140381

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140381
Availability details for
Quantity
  • Availability
  • Add to Cart