ENA413166522 Enamine

2-(chloromethyl)-6-methoxy-1,3-benzothiazole

  • MDL number MFCD11217502
  • Cas Number : 1056639-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141874

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141874
Availability details for
Quantity
  • Availability
  • Add to Cart