ENA413166531 Enamine

2-cyclohexyl-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD11845269
  • Cas Number : 66493-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141906

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141906
Availability details for
Quantity
  • Availability
  • Add to Cart