ENA413166532 Enamine

4-amino-2,3,6-trimethylphenol

  • MDL number MFCD19300989
  • Cas Number : 17959-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133...135
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141908

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141908
Availability details for
Quantity
  • Availability
  • Add to Cart