ENA413166537 Enamine

1-cyclopropyl-8-methyl-1,2,3,4-tetrahydroquinoxaline

  • MDL number MFCD24229044
  • Cas Number : 1553271-94-5
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-cyclopropyl-8-methyl-1,2,3,4-tetrahydroquinoxaline
Inchi Code 1S/C12H16N2/c1-9-3-2-4-11-12(9)14(8-7-13-11)10-5-6-10/h2-4,10,13H,5-8H2,1H3
Inchi Key VSJIFSLMUQAXHK-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-141920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141920
Availability details for
Quantity
  • Availability
  • Add to Cart