ENA413166546 Enamine

3-(6-methylpyridin-3-yl)propanal

  • MDL number MFCD20336883
  • Cas Number : 1342446-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141930

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141930
Availability details for
Quantity
  • Availability
  • Add to Cart