ENA413166549 Enamine

2-(chlorosulfonyl)benzene-1-sulfonyl fluoride

  • MDL number MFCD00065074
  • Cas Number : 30672-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88...90
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141937

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141937
Availability details for
Quantity
  • Availability
  • Add to Cart