ENA413166554 Enamine

potassium 5-(methoxymethyl)-1,2,4-oxadiazole-3-carboxylate

  • MDL number MFCD26936231
  • Cas Number : 1607281-07-1
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name potassium 5-(methoxymethyl)-1,2,4-oxadiazole-3-carboxylate
Inchi Code 1S/C5H6N2O4.K/c1-10-2-3-6-4(5(8)9)7-11-3;/h2H2,1H3,(H,8,9);/q;+1/p-1
Inchi Key DGAQZTLUJLLRDN-UHFFFAOYSA-M

Description

Purity 95
Salt Data [K+]
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-141943

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-141943
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart