ENA413166563 Enamine

N-ethyl-N'-(propan-2-ylidene)(tert-butoxy)carbohydrazide

  • MDL number MFCD10699981
  • Cas Number : 561067-07-0
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-141977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141977
Availability details for
Quantity
  • Availability
  • Add to Cart