ENA413166567 Enamine

tert-butyl 4-[(1,3-thiazol-2-yl)amino]piperidine-1-carboxylate

  • MDL number MFCD22231811
  • Cas Number : 849546-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141983

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141983
Availability details for
Quantity
  • Availability
  • Add to Cart