ENA413166568 Enamine

4-bromo-1-(oxolan-3-yl)-1H-pyrazole

  • MDL number MFCD18366437
  • Cas Number : 1040377-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141987
Availability details for
Quantity
  • Availability
  • Add to Cart