ENA413166573 Enamine

2-chloro-6-(piperidin-4-yloxy)pyrazine hydrochloride

  • MDL number MFCD13561256
  • Cas Number : 1220036-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141992
Availability details for
Quantity
  • Availability
  • Add to Cart