ENA413166575 Enamine

1-(4-nitrophenyl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD00592069
  • Cas Number : 14213-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139783

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139783
Availability details for
Quantity
  • Availability
  • Add to Cart