ENA413166576 Enamine

1-ethyl-4-fluoro-2-[(1H-imidazol-1-yl)methyl]-1H-1,3-benzodiazole

  • MDL number MFCD26936236
  • Cas Number : 1311923-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141995
Availability details for
Quantity
  • Availability
  • Add to Cart