ENA413166588 Enamine

3-methyl-2,3,4,5-tetrahydro-1,4-benzothiazepin-5-one

  • MDL number MFCD24576314
  • Cas Number : 24187-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142025
Availability details for
Quantity
  • Availability
  • Add to Cart