ENA413166595 Enamine

1-(6-chloropyrazin-2-yl)azepane

  • MDL number MFCD13561858
  • Cas Number : 1138220-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140402

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140402
Availability details for
Quantity
  • Availability
  • Add to Cart