ENA413166596 Enamine

3-tert-butyl-1-[(pyridin-2-yl)methyl]-1H-pyrazol-5-amine

  • MDL number MFCD26936242
  • Cas Number : 1803580-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-142054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142054
Availability details for
Quantity
  • Availability
  • Add to Cart