ENA413166606 Enamine

1-(3,4-difluorophenyl)-2-methylprop-2-en-1-amine hydrochloride

  • MDL number MFCD26936248
  • Cas Number : 1803610-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260...262
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142098

Safety Information

msds https://www.enaminestore.com/catalog/EN300-142098
Availability details for
Quantity
  • Availability
  • Add to Cart