ENA413166610 Enamine

5-(methylsulfanyl)pentan-1-amine

  • MDL number MFCD14685303
  • Cas Number : 55021-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-142102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142102
Availability details for
Quantity
  • Availability
  • Add to Cart