ENA413166619 Enamine

2-oxaspiro[4.4]nonan-1-one

  • MDL number MFCD25964832
  • Cas Number : 5732-98-9
  • Molecular Weight : 140.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-oxaspiro[4.4]nonan-1-one
Inchi Code 1S/C8H12O2/c9-7-8(5-6-10-7)3-1-2-4-8/h1-6H2
Inchi Key PBZONIMHAVOWHW-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-142118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-142118
Availability details for
Quantity
  • Availability
  • Add to Cart